Human cleaved C-terminal GSDMD Antibody Pair - BSA and Azide Free
Catalog # ABCAAB314572-10X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:cleaved C-terminal GSDMD
- Description:Human cleaved C-terminal GSDMD antibody pair - BSA and azide free
- Detection range:15,6 - 1000 pg/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Human cleaved C-terminal GSDMD antibody pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human cleaved C-terminal GSDMD.