Mouse/Rat proANP Antibody Pair - BSA and Azide Free
Catalog # ABCAAB270357-10X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:proANP
- Description:Mouse/Rat proANP antibody pair - BSA and azide free
- Detection range:125 - 8000 pg/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Mouse/Rat proANP antibody pair - BSA and Azide free.