Order Entry
France
ContactUsLinkComponent
Mouse Transferrin Receptor Antibody Pair - BSA and Azide free
Mouse Transferrin Receptor Antibody Pair - BSA and Azide free
Catalog # ABCAAB256631-10X96
Supplier:  Abcam
CAS Number:  
undefined
Mouse Transferrin Receptor Antibody Pair - BSA and Azide free
Catalog # ABCAAB256631-10X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Mouse Transferrin Receptor antibody pair - BSA and Azide free
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    10×96 tests

Specifications

About this item

Mouse Transferrin Receptor Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse Transferrin Receptor.