Mouse YKL-40/CHI3L1 Antibody Pair - BSA and Azide Free
Catalog # ABCAAB244154-10X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:YKL-40/CHI3L1
- Description:Mouse YKL-40 / CHI3L1 antibody pair - BSA and azide free
- Detection range:54,69 - 3500 pg/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Mouse YKL-40 / CHI3L1 antibody pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse YKL-40 / CHI3L1.