Mouse Tissue Factor Antibody Pair - BSA and Azide free
Catalog # ABCAAB241762-10X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Tissue Factor antibody pair - BSA and Azide free
- Storage temperature:+4 ℃
- Tests per kit:10×96 tests
Specifications
About this item
Mouse tissue Factor Antibody Pair - BSA and Azide free.