Human Surfactant protein D/SP-D Antibody Pair - BSA and Azide free
Catalog # ABCAAB244174-10X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Surfactant protein D/SP-D antibody pair - BSA and Azide free
- Storage temperature:+4 ℃
- Tests per kit:10×96 tests
Specifications
About this item
Human Surfactant protein D/SP-D Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Surfactant protein D/SP-D.