Mouse EMMPRIN Antibody Pair - BSA and Azide free
Catalog # ABCAAB241741-10X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse EMMPRIN Antibody Pair - BSA and Azide free
- Tests per kit:10×96 tests
Specifications
About this item
Mouse EMMPRIN Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse EMMPRIN.