Mouse/Rat Heme Oxygenase 1 Antibody Pair - BSA and Azide free
Catalog # ABCAAB241763
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse/Rat Heme Oxygenase 1 Antibody Pair - BSA and Azide free
- Tests per kit:10×96 tests
Specifications
About this item
Mouse/Rat Heme Oxygenase 1 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse/Rat Heme Oxygenase 1.