cis-9-Tricosene (muscalure)
Catalog # EHERC15350000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
- Environmentally Preferable:
Specifications
About this item
Synonyms: (Z)-9-Tricosene
:
:322.62 g/mol
EINECS:248-505-7
CAS Number:27519-02-4
Formula:C₂₃H₄₆
MDL Number:MFCD00008988
Boiling Pt:300
Density:0.806 g/cm³ (20 °C)
Merck Index:12,06288