Incinerating dishes
Alsint 99,7.
- Curved suvertical surfaces for easy removal of sample
- Good thermal shock resistance due to high thermal conductivity
- Resistant to chemical attack, e.g. hydrogen and other reducing gases
Frequently Bought Together
Alsint 99,7.