Order Entry
France
Orders LinkContactUsLinkComponent
Decade boxes
Decade boxes
Supplier:   ELC
Decade boxes
Supplier:  ELC
Certificates
Decade boxes
7 Options Available Below
Product Details & Documents
Decade boxes for testing of electronic circuits with different resistance, capacitance or inductance.
  • Robust switches with metal frames and gold contacts
  • Safety sockets
  • Front panel fits a standard sub rack
  • Complies with EN 61010-1
Capacitance decade box: Max voltage: 300 V DC or 230 V AC; high accuracy 1% and low temperature coefficient; discharge button.
Inductance decade box: Accuracy: 5%; inductors wound round a ferrite pot core - Q factor between 55 and 100; max voltage 150 V AC/DC or 250 V AC/DC with earth connected.
Resistance decade boxes: The DR04 - DR07 boxes use metal film resistors offering 0,5% accuracy and temperature coefficientof 50 ppm/°C; the DR08 box uses thick film resistors offering 1% accuracy and temperature coefficientof 100 ppm/°C.
Product Family Options
Capacitance decade box
Product Information
  • Availability
    Price
  • 139-0397
    5 decade capacitance box, DC 05, 100 pF to 11,111 µF
    5 decade capacitance box, DC 05, 100 pF to 11,111 µF
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 424,41 €
    424,41 €
    1 * 1 Pce
       424,41 €
Inductance decade box
Product Information
  • Availability
    Price
  • 139-0421
    7 decade inductance box, DL 07, 1 µH to 11,111110 H
    7 decade inductance box, DL 07, 1 µH to 11,111110 H
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 403,04 €
    403,04 €
    1 * 1 Pce
       403,04 €
Resistance decade boxes
Product Information
  • Availability
    Price
  • 139-0398
    4 decade resistance box, DR 04, 1 Ω to 11.110 kΩ
    4 decade resistance box, DR 04, 1 Ω to 11.110 kΩ
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 186,25 €
    186,25 €
    1 * 1 Pce
       186,25 €
  • 139-0422
    5 decade resistance box, DR 05, 1 Ω to 111.110 kΩ
    5 decade resistance box, DR 05, 1 Ω to 111.110 kΩ
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 212,21 €
    212,21 €
    1 * 1 Pce
       212,21 €
  • 139-0423
    6 decade resistance box, DR 06, 1 Ω to 1,111110 MΩ
    6 decade resistance box, DR 06, 1 Ω to 1,111110 MΩ
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 238,16 €
    238,16 €
    1 * 1 Pce
       238,16 €
  • 139-0399
    7 decade resistance box, DR 07, 1 Ω to 11.111110 MΩ
    7 decade resistance box, DR 07, 1 Ω to 11.111110 MΩ
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 273,28 €
    273,28 €
    1 * 1 Pce
       273,28 €
  • 139-0401
    8 decade resistance box, DR 08, 0.1 Ω to 11.111111 MΩ
    8 decade resistance box, DR 08, 0.1 Ω to 11.111111 MΩ
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 297,71 €
    297,71 €
    1 * 1 Pce
       297,71 €