2-Iodoadenosine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
About this item
Synonyms: (2R,3R,4S,5R)-2-(6-Amino-2-iodo-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
- CAS Number:35109-88-7
- Formula:C₁₀H₁₂IN₅O₄
- MDL Number:MFCD01320413
- Molecular Weight:393.14 g/mol
- Melting Pt:144°C
3 Options Available Below
Product Details & Documents
Synonyms: (2R,3R,4S,5R)-2-(6-Amino-2-iodo-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
- CAS Number:35109-88-7
- Formula:C₁₀H₁₂IN₅O₄
- MDL Number:MFCD01320413
- Molecular Weight:393.14 g/mol
- Melting Pt:144°C
Specifications for Product Family
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



