Miniature luer connectors, BOLA
Connector made of PTFE for transition from Luer connection to female thread UNF ¼" 28 G.
- Temperature resistance from -200 °C to +250 °C
- Universal chemical resistance
Frequently Bought Together
Connector made of PTFE for transition from Luer connection to female thread UNF ¼" 28 G.