Order Entry
France
ContactUsLinkComponent
Capillary columns, Elite-Guard and Elite-Siltek® for autosystem
Capillary columns, Elite-Guard and Elite-Siltek® for autosystem
  PERCN9316608
Capillary columns, Elite-Guard and Elite-Siltek® for autosystem
  PERCN9316608
  • Column Format:
    Capillary
  • Ø int.:
    0,32 mm
  • Description:
    Elite-Siltek Guard
  • Length:
    5 m
  • Seq:
    08
  • Pk:
    1
These guard columns lengthen the life of the capillary column and improve the analyte focusing.
  • Increase the life of the capillary column and performance
  • Maintains retention time and resolution on capillary column
  • Traps non-volatile particles
Both the intermediate-polarity deactivated guard columns and the Siltek® deactivated guard columns are important to match the polarity of the solvent and the polarity of the surface deactivation. Temperature range is 325 °C for Elite-Guardand 380 °C for Elite-Siltek® guard.