
1,4,7-Triazacyclononane trihydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:58966-93-1
:
Formula:C₆H₁₈Cl₃N₃
MDL Number:MFCD00074887
:238.5900 g/mol
Melting Pt:285…290 °C
Storage Temperature:Ambient
UN:3259
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Off-white to yellow Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥96.0 %
Specification Test Results
Frequently Bought Together