Potassium permanganate saturated solution in water
Catalog # PRLSMC306820.0500
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
About this item
Synonyms: Permanganic acid potassium salt
:
:158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
UN:3082
ADR:9,III
:GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364