Order Entry
France
ContactUsLinkComponent
Nissel tubes
Nissel tubes
Catalog # HECH41145010
undefined
Nissel tubes
Catalog # HECH41145010
Specifications
  • Description:
    Tube Nissel (3 ml), with blue graduation, for the volumetric determination of fibrin
  • Pk:
    1
Specifications
About this item
Glass.
  • Fused blue graduation
For the volumetric determination of fibrin. With directions for use - "heat fibrin according to Schulz".