Order Entry
France
ContactUsLinkComponent
(2R,3R)-(-)-2,3-O-Isopropylidene-D-threitol for synthesis
(2R,3R)-(-)-2,3-O-Isopropylidene-D-threitol for synthesis
Catalog # 8.18636.0001
Supplier:  Merck
CAS Number:  
(2R,3R)-(-)-2,3-O-Isopropylidene-D-threitol for synthesis
Catalog # 8.18636.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    46 - 49 °C
  • Molar Mass:
    162.18 g/mol
  • Boiling Point:
    110 - 120 °C (1 hPa)
  • Flash Point:
    1.4574 (20 °C, 589 nm)
  • Density:
    1.15 g/cm3 (20 °C)
  • Synonyms:
    (4R,5R)-(-)-4,5-Bis(hydroxymethyl)-2,2-dimethyl-1,3-dioxolane
  • Hill Formula:
    C7H14O4

Specification Test Results

About this item

 :
 :162.18 g/mol
EINECS:277-391-1
CAS Number:73346-74-4
Formula:C₇H₁₄O₄
MDL Number:MFCD00009761