Propanephosphonic acid cyclic anhydride 50% in DMF, T3P® for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:68957-94-8
:
Formula:C₉H₂₁O₆P₃
MDL Number:MFCD00006583
:318.19 g/mol
ADR:8 (3),II
:M-Clarity™ quality level = MQ200
EINECS:422-210-5
UN:2920
:GHS02
:GHS05
:GHS08
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Boiling Point:153 °C (1013 hPa) (Dimethylformamide)
- Flash Point:58 °C (Dimethylformamide)
Specification Test Results
Frequently Bought Together