


Propanephosphonic acid cyclic anhydride 50% in ethyl acetate, Sigma-Aldrich®
: Merck
:68957-94-8
:
:C₉H₂₁O₆P₃
:MFCD00006583
:318.19 g/mol
:3 (8),II
:M-Clarity™ quality level = MQ200
:422-210-5
:2924
:GHS02
:GHS05
:GHS07
:Refrigerator
- Boiling Point:77 °C (1013 hPa) ethyl acetate
- Vapor Pressure:97 hPa (20 °C)
- Flash Point:-4 °C ethyl acetate
- Density:1.08 g/cm3 (20 °C)
Frequently Bought Together