1,1-Diphenylhydrazine hydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:530-47-2
:
Formula:C₁₂H₁₃ClN₂
MDL Number:MFCD00012950
:220.70 g/mol
Merck Index:14,3325
Melting Pt:∼ 163 °C (dec.)
EINECS:208-481-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together