Order Entry
France
ContactUsLinkComponent
Nicotinic acid hydrazide 97%
Irritant
Health
Nicotinic acid hydrazide 97%
Certificates
undefined
Irritant
Health
Nicotinic acid hydrazide 97%
Certificates

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    160 to 163 °C
  • Assay:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together