Order Entry
France
ContactUsLinkComponent
Magnesium acetate tetrahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Magnesium acetate tetrahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2424-01
Supplier:  Avantor
CAS Number:  
undefined
Magnesium acetate tetrahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2424-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    98.0 - 102.0 %
  • Assay ((CH₃COO)₂Mg·4H₂O):
    98.0 - 102.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    6.5 - 8.5
  • Chloride (Cl):
    ≤0.001 %
  • Sulfate (SO₄):
    ≤0.005 %
  • Barium (Ba):
    ≤0.001 %
  • Calcium (Ca):
    ≤0.01 %
  • Manganese (Mn):
    ≤0.001 %
  • Potassium (K):
    ≤0.005 %
  • Sodium (Na):
    ≤0.005 %
  • Strontium (Sr):
    ≤0.005 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

About this item

 :
 :214.46 g/mol
Merck Index:13,05676
Melting Pt:80 °C
Density:1.454 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-554-9
CAS Number:16674-78-5
Formula:(H₃CCOO)₂Mg·4H₂O
MDL Number:MFCD00149214