Graduated pipettes, Class AS
Supplier: witeg Labortechnik
Jump to:
About this item
Graduated pipettes, class AS, conformity certified.
- For complete swift delivery
- With amber stain main point ring-graduation
- Tempered delivery tips offering a wear resistance
- With construction in the suction to hold cotton plug
- Waiting time 3 s
Certifications: DIN 12697
2 Options Available Below
Product Details & Documents
Graduated pipettes, class AS, conformity certified.
- For complete swift delivery
- With amber stain main point ring-graduation
- Tempered delivery tips offering a wear resistance
- With construction in the suction to hold cotton plug
- Waiting time 3 s
Certifications: DIN 12697
Product Family Options
Product Information