Order Entry
France
ContactUsLinkComponent
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)
Catalog # MOLE16832467-5MG
Supplier:  Molekula
CAS Number:  
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)
Catalog # MOLE16832467-5MG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :308.4 · 36.5 g/mol
CAS Number:65391-42-6
Formula:C₁₆H₂₅ClN₂O₄
MDL Number:MFCD00058004