Order Entry
France
Orders LinkContactUsLinkComponent
 

Adenosine

Glass bottle for solids, 1 g
Catalog # ROTH2322.3
Marketsource
Supplier:   Roth Carl
Adenosine
Catalog # ROTH2322.3
Supplier:  Roth Carl
Marketsource
CAS Number:  58-61-7
Adenosine

Glass bottle for solids, 1 g

CAS Number:  58-61-7
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  • CAS Number:58-61-7
  • Formula:C₁₀H₁₃N₅O₄
  • MDL Number:MFCD00005752
  • Molecular Weight:267.24 g/mol
  • Storage Temperature:Réfrigérateur
  • EINECS:200-389-9
Specifications

Specifications

  • Type de conditionnement:
    Glass bottle for solids
  • Cdt:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.