Adenosine ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 58-61-7 Molecular Formula: C10H13N5O4 Formula Weight: 267.25 Storage Temperature: 2°C to 8°C Physical Form: Powder Odour: Odourless Melting Point: 234-237° Beilstein Reference: 93029 Merck Reference: 14,153 Optical Rotation: -70° (c=2 in 5% NaOH) MDL No.: MFCD00005752
Synonyms: 9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
- CAS Number:58-61-7
- Formula:C₁₀H₁₃N₅O₄
- MDL Number:MFCD00005752
- Molecular Weight:267.24 g/mol
- Merck Index:14,153
- Melting Pt:234…237 °C
- Storage Temperature:Refrigerator
- EINECS:200-389-9
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 58-61-7 Molecular Formula: C10H13N5O4 Formula Weight: 267.25 Storage Temperature: 2°C to 8°C Physical Form: Powder Odour: Odourless Melting Point: 234-237° Beilstein Reference: 93029 Merck Reference: 14,153 Optical Rotation: -70° (c=2 in 5% NaOH) MDL No.: MFCD00005752
Synonyms: 9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
- CAS Number:58-61-7
- Formula:C₁₀H₁₃N₅O₄
- MDL Number:MFCD00005752
- Molecular Weight:267.24 g/mol
- Merck Index:14,153
- Melting Pt:234…237 °C
- Storage Temperature:Refrigerator
- EINECS:200-389-9



