Order Entry
France
ContactUsLinkComponent
3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl)
3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl)
Catalog # 281751-10
CAS Number:  
3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl)
Catalog # 281751-10
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :360.11 g/mol
EINECS:231-018-9
CAS Number:7411-49-6
Formula:C₁₂H₁₈Cl₄N₄
MDL Number:MFCD00012969