Order Entry
France
ContactUsLinkComponent
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA)
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA)
Catalog # MOLE10783994-5G
Supplier:  Molekula
CAS Number:  
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA)
Catalog # MOLE10783994-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :241.33 g/mol
CAS Number:77377-52-7
Formula:C₉H₁₈F₃NOSi
MDL Number:MFCD00009671