Order Entry
France
ContactUsLinkComponent
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) 98%
Irritant
Flammable
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) 98%
Certificates
undefined
Irritant
Flammable
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) 98%
Certificates

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4010 to 1.4030 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together