Order Entry
France
ContactUsLinkComponent
Methyl green zinc chloride salt 1% in aqueous solution
Methyl green zinc chloride salt 1% in aqueous solution
Catalog # TREV4800-30-18
Supplier:  TREVIGEN
CAS Number:  
Methyl green zinc chloride salt 1% in aqueous solution
Catalog # TREV4800-30-18
Supplier:  TREVIGEN
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :653.24 g/mol
EINECS:230-415-4
CAS Number:7114-03-6
Formula:C₂₇H₃₅BrClN₃·xZnCl₂
MDL Number:MFCD00151094