(2-Nitrophenyl)-β-D-galactopyranoside (ONPG) for synthesis, Sigma-Aldrich®
Catalog # 8.41747.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
Specification Test Results
- Molar Mass:301.25 g/mol
- Hill Formula:C12H15NO8
Specification Test Results
About this item
Synonyms: o-Nitrophenyl-β-D-galactoside, 2-Nitrophenyl β-D-Galactopyranoside, O-Nitrophenyl-Β-D-Galactopyranoside, ONPG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol
:
:301.25 g/mol
Storage Temperature:Freezer
EINECS:206-716-1
CAS Number:369-07-3
Formula:C₁₂H₁₅NO₈
MDL Number:MFCD00063255
:M-Clarity™ quality level = MQ200