


Methyl green zinc chloride salt <0,5% crystal violet 80% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:7114-03-6
:
Formula:C₂₇H₃₅BrClN₃·xZnCl₂
MDL Number:MFCD00151094
:653.24 g/mol
Melting Pt:>300 °C (lit.)
:M-Clarity™ quality level = MQ200
EINECS:230-415-4
:GHS07
:GHS05
:GHS09
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Form); Powder or Crystalline Powder:Conforms to Requirements
- Infrared spectrum:Conforms to Structure
- Wavelength, c = 0.005 g/L; Water:629 - 635 nm
- Extinction Coefficient:≥50000
- Wavelength:420 - 426 nm
- Carbon:37.2 - 62.0 %
- Nitrogen:4.8 - 8.0 %
- ICP Major Analysis, Confirms Zinc Component:Confirmed
- Purity (HPLC):≥75.0 %
- Crystal Violet:≤0.5 %
- Solubility (Colour), c = 1 mg/ml; Water:Dark Blue to Very Dark Blue and Dark Blue-Green to Very Dark Blue-Green
Specification Test Results
Frequently Bought Together