Order Entry
France
ContactUsLinkComponent
6-Azauracil for synthesis, Sigma-Aldrich®
6-Azauracil for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
6-Azauracil for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Melting Point:
    274 - 275 °C
  • Molar Mass:
    113.07 g/mol
  • Synonyms:
    1,2,4-Triazine-3,5(2H,4H)-dione
  • Hill Formula:
    C3H3N3O2

Specification Test Results

Related Information

Frequently Bought Together