Order Entry
France
ContactUsLinkComponent
o-Cresolphthalein, Sigma-Aldrich®
o-Cresolphthalein, Sigma-Aldrich®
Catalog # C85778-10G
CAS Number:  
undefined
o-Cresolphthalein, Sigma-Aldrich®
Catalog # C85778-10G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :346.39 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:209-881-8
CAS Number:596-27-0
Formula:C₂₂H₁₈O₄
MDL Number:MFCD00005912