Racks for chest freezers
Supplier: GLW
Certificates
About this item
These freezer rack are commonly used for all chest freezers and available in various exterior heights.
- Made of stainless steel
- Can accomodate 18 boxes
- With one folding handle on top
Ordering information: Stainless steel locking rod has to be ordered separately.
Frequently Bought Together