Pallets, gas cylinders
About this item
These pallets are used for the safe, economic and effortless transport of 1 to 8 gas cylinders with hot-dip galvanised surface.
- Sturdy base frame with fortlift pockets
- Gas cylinders can be secured individually with chains
- Lateral tilt limiter
- Hooks for hoses and screw caps
Inner dimensions of fortlift pockets (W×H): 170×70 mm.
Inner distance between pockets: 510 mm.
Certifications: According to EN ISO 1461
2 Options Available Below
Product Details & Documents
These pallets are used for the safe, economic and effortless transport of 1 to 8 gas cylinders with hot-dip galvanised surface.
- Sturdy base frame with fortlift pockets
- Gas cylinders can be secured individually with chains
- Lateral tilt limiter
- Hooks for hoses and screw caps
Inner dimensions of fortlift pockets (W×H): 170×70 mm.
Inner distance between pockets: 510 mm.
Certifications: According to EN ISO 1461
Product Family Options
Product Information
- W×D×HAvailabilityPrice
- DUEH7.00.800Transport pallet for max. 4× Ø 250 mm gas cylinders, load capacity 350 kgTransport pallet for max. 4× Ø 250 mm gas cylinders, load capacity 350 kg600×860×1000 mmLog in for availability1 * 1 Pce394,00 €
- DUEH7.00.810Transport pallet for max. 8× Ø 250 mm gas cylinders, load capacity 700 kgTransport pallet for max. 8× Ø 250 mm gas cylinders, load capacity 700 kg1200×860×1000 mmLog in for availability1 * 1 Pce541,00 €