Pump Tubing, Silicone
Supplier: Heidolph Instruments GmbH & Co.KG
Certificates
About this item
Polydimethyl siloxane with siliceous earth and silicone additives; translucent white, excellent resistance to initial pressure.
- Extremely smooth interior prevents bacterial growth
- Biocompatible, minimal adsorption and absorption
- Best flow properties, high temperature stability
- Absolutely inert, softener-free
Platinum-coated silicone hose for use in pharmaceuticals and biology.
Frequently Bought Together