Order Entry
France
Orders LinkContactUsLinkComponent
Hydrochloric acid 5%

Hydrochloric acid 5%

Corrosive
Supplier:   PROLABO CMP
Hydrochloric acid 5%
Supplier:  PROLABO CMP
Certificates
Hydrochloric acid 5%

2 Options Available Below

Product Details & Documents
Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Boiling Pt:∼100 °C (1013 hPa)
  • Density:1.02…1.05 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 42,20 €
    42,20 €
    1 * 1 L
       42,20 €

  • 20 L
    Log in for availability
    Log in for availability
    1 * 20 L
    1 * 20 L - 222,00 €
    222,00 €
    1 * 20 L
       222,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.