Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrochemistry Meters
pH and Multi-Parameter Meters
Accessories for pH/°C meter, handheld, 206
Accessories for pH/°C meter, handheld, 206
Accessories for pH/°C meter, handheld, 206
Catalog # TEST0554.2067
Supplier:
TESTO
Expand image
Accessories for pH/°C meter, handheld, 206
Catalog # TEST0554.2067
Supplier:
TESTO
Specifications
Description:
Storage cap for testo 206, with KCI gel filling
Pk:
1
Specifications
About this item
Product Details
Product Details