Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dragendorff's reagent
Dragendorff's reagent
Dragendorff's reagent
PRLSMC302342.0100
:
PROLABO CMP
:
Dragendorff's reagent
PRLSMC302342.0100
:
PROLABO CMP
:
Pack type:
Plastic bottle
Pk:
100 mL
Seq:
2440
:
:
77 °C (1013 hPa)
:
1.2 g/cm³ (20 °C)
:
–4 °C
:
Ambient
:
39775-75-2
:
MFCD00283106