Order Entry
France
ContactUsLinkComponent
Tube rack, for water bath, SP Bel-Art
  BELAF188820000
undefined
Tube rack, for water bath, SP Bel-Art
  BELAF188820000
  • For tubes:
    1,5 − 15 ml
  • No. of Holes:
    6
  • Description:
    Rack for water bath
  • Material:
    PP
  • Environmentally Preferable:
  • Pk:
    1
Height-adjustable racks securely hold up to six tubes or vials at any desired depth in a water bath or beaker. The rack has a flexible grip that easily retains mixed test tubes of varying weight and volume from 1,5 ml to 15 ml.
  • Made of PP
  • Adjustable height prevents submersion problems
  • Compact footprint
  • Stable base
The compact yet stable footprint saves bench and bath space and lifts in and out of a bath without splashing.