Silver standard dissolved in oil, 1,000 µg/g Ag in Base Oil 75 for wear metal analysis
Catalog # JOBIORG-AG8-2Z
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 µg/g
- Matrix:oil
- Pack type:Plastic bottle for solids
- Pk:50 g
- Description:Standard for wear metal analysis
- Element:Silver
- Symbol:Ag