Calcium standard dissolved in oil, 5,000 µg/g Ca in Base Oil 75 for wear metal analysis
Référence JOBIORG-CA8-4Z
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:5000 µg/g
- Matrix:Base Oil 75
- Pack type:Glass bottle for solids
- Pk:50 g
- Element:Calcium
- Symbol:Ca