Order Entry
France
ContactUsLinkComponent
Boron standard dissolved in oil, 5,000 µg/g B in Base Oil 75 for wear metal analysis
Boron standard dissolved in oil, 5,000 µg/g B in Base Oil 75 for wear metal analysis
  JOBIORG-B8-4Z
 :  
Boron standard dissolved in oil, 5,000 µg/g B in Base Oil 75 for wear metal analysis
  JOBIORG-B8-4Z
 :  

 

  • Concentration:
    5000 µg/g
  • Matrix:
    Base Oil 75
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Element:
    Boron
  • Symbol:
    B