Boron standard dissolved in oil, 1,000 µg/g B in Base Oil 75 for wear metal analysis
Catalog # JOBIORG-B8-2Z
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 µg/g
- Matrix:Base Oil 75
- Pack type:Glass bottle for solids
- Pk:50 g
- Element:Boron
- Symbol:B