Order Entry
France
ContactUsLinkComponent
Bismuth standard dissolved in oil, 1,000 µg/g Bi in Base Oil 75 for wear metal analysis
Bismuth standard dissolved in oil, 1,000 µg/g Bi in Base Oil 75 for wear metal analysis
Catalog # JOBIORG-BI8-2Z
CAS Number:  
Bismuth standard dissolved in oil, 1,000 µg/g Bi in Base Oil 75 for wear metal analysis
Catalog # JOBIORG-BI8-2Z
CAS Number:  

Specifications

  • Concentration:
    1000 µg/g
  • Matrix:
    Base Oil 75
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Element:
    Bismuth
  • Symbol:
    Bi

Specifications

About this item