Barium standard dissolved in oil, 5,000 µg/g Ba in Base Oil 75 for wear metal analysis
Catalog # JOBIORG-BA8-4Z
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Base Oil 75
- Pack type:Glass bottle for solids
- Pk:50 g
- Element:Barium
- Symbol:Ba