Order Entry
France
ContactUsLinkComponent
Organic reference standard, 4-Bromophenyl phenyl ether (BDE No. 3) 50 µg/ml in n-nonane, CIL
Organic reference standard, 4-Bromophenyl phenyl ether (BDE No. 3) 50 µg/ml in n-nonane, CIL
Catalog # PROOCIL-BDE-3-CS
CAS Number:  
Organic reference standard, 4-Bromophenyl phenyl ether (BDE No. 3) 50 µg/ml in n-nonane, CIL
Catalog # PROOCIL-BDE-3-CS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    101-55-3
  • Description:
    4-Bromophenyl phenyl ether (BDE No. 3) 50 µg/ml in n-nonane
  • Formula:
    C₁₂H₉BrO
  • Description:
    4-Bromophenyl phenyl ether (BDE No. 3)

Specifications

About this item