Herbicide reference standard, Linuron (N-(3,4-Dichlorophenyl)-N'-methoxy-N'-methylurea) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA14640000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:330-55-2
- Description:Linuron (N-(3,4-Dichlorophenyl)-N'-methoxy-N'-methylurea) 100 µg/ml in acetonitrile
- Formula:C₉H₁₀Cl₂N₂O₂
- Description:Linuron (N-(3,4-Dichlorophenyl)-N'-methoxy-N'-methylurea)